2-Amino-6-methylbenzonitrile 98%
Catalog # APOSOR926664-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:132.17
Storage Temperature:Ambient
CAS Number:56043-01-7
Formula:C₈H₈N₂
MDL Number:MFCD00058686