0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
tert-Butyl-2-oxoazetidine-1-carboxylate
tert-Butyl-2-oxoazetidine-1-carboxylate 95%
tert-Butyl-2-oxoazetidine-1-carboxylate 95%
tert-Butyl-2-oxoazetidine-1-carboxylate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
tert-Butyl-2-oxoazetidine-1-carboxylate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1140510-99-1
:
Formula:
C₈H₁₃NO₃
MDL Number:
MFCD08751345
:
171.2 g/mol
Storage Temperature:
Refrigerator