Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium pantothenate
Calcium pantothenate 95%
Calcium pantothenate 95%
Calcium pantothenate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Calcium pantothenate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
137-08-6
:
Formula:
C₁₈H₃₂CaN₂O₁₀
MDL Number:
MFCD00002766
:
476.54 g/mol
Melting Pt:
190 °C
:
GHS07
Storage Temperature:
Ambient