Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyrocatechol 99%

Pyrocatechol 99%

New Product
Supplier:   Apollo Scientific
Pyrocatechol 99%
Supplier:  Apollo Scientific
New Product
Certificates
Pyrocatechol 99%

4 Options Available Below

Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Boiling Pt:245 °C
  • Melting Pt:103…106 °C
  • Flash Pt:131 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 26,10 €
    26,10 €
    1 * 5 g
       26,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 31,90 €
    31,90 €
    1 * 10 g
       31,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 39,10 €
    39,10 €
    1 * 25 g
       39,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 47,80 €
    47,80 €
    1 * 50 g
       47,80 €

    Specifications:
    New Product
    New Product