Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Phenyl-1-propanol
3-Phenyl-1-propanol 95%
3-Phenyl-1-propanol 95%
3-Phenyl-1-propanol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Phenyl-1-propanol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
122-97-4
:
Formula:
(C₆H₅)CH₂CH₂CH₂OH
MDL Number:
MFCD00002950
:
136.19 g/mol
Storage Temperature:
Ambient