Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,5,6-Tetramethylbenzoic acid
2,3,5,6-Tetramethylbenzoic acid 95%
2,3,5,6-Tetramethylbenzoic acid 95%
2,3,5,6-Tetramethylbenzoic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3,5,6-Tetramethylbenzoic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2604-45-7
:
Formula:
C₁₁H₁₄O₂
:
178.23 g/mol
Storage Temperature:
Ambient