(4-Methoxyphenyl)acetic acid ethyl ester 95%
Catalog # APOSOR920393-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:194.23 g/mol
Storage Temperature:Ambient
CAS Number:14062-18-1
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00040760