Order Entry
Germany
ContactUsLinkComponent
4-sec-Butylcyclohexylamine (cis- and trans- mixture) 95%
4-sec-Butylcyclohexylamine (cis- and trans- mixture) 95%
Catalog # APOSOR920000-5G
Supplier:  Apollo Scientific
CAS Number:  
4-sec-Butylcyclohexylamine (cis- and trans- mixture) 95%
Catalog # APOSOR920000-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :155.29 g/mol
Storage Temperature:Ambient
CAS Number:25834-93-9
Formula:C₁₀H₂₁N