N-sec-Butylpropylamine 95%
Catalog # APOSOR919285-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:115.22 g/mol
Storage Temperature:Ambient
CAS Number:39190-67-5
Formula:C₇H₁₇N
MDL Number:MFCD00059398