3-Methyl-4-heptanone 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15726-15-5
:
Formula:C₈H₁₆O
MDL Number:MFCD00048721
:128.21 g/mol
Boiling Pt:156 °C
Density:0.82 g/cm³ (25 °C)
Flash Pt:38 °C
Storage Temperature:Ambient