Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iproniazid
Iproniazid 95%
Iproniazid 95%
Iproniazid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Iproniazid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
54-92-2
:
Formula:
C₉H₁₃N₃O
MDL Number:
MFCD00153819
:
179.22 g/mol
Storage Temperature:
Ambient