Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Pentylcyclohexanone 98%

New Product
Supplier:   Apollo Scientific
4-Pentylcyclohexanone 98%
Supplier:  Apollo Scientific
New Product
Certificates
4-Pentylcyclohexanone 98%

6 Options Available Below

Product Details & Documents
  • CAS Number:61203-83-6
  • Formula:C₁₁H₂₀O
  • MDL Number:MFCD00059581
  • Molecular Weight:168.28 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,00 €
    29,00 €
    1 * 5 g
       29,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 82,50 €
    82,50 €
    1 * 25 g
       82,50 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 194,00 €
    194,00 €
    1 * 100 g
       194,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 286,00 €
    286,00 €
    1 * 500 g
       286,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 400,00 €
    1 400,00 €
    1 * 2,5 kg
       1 400,00 €

    Specifications:
    New Product
    New Product