Order Entry
Germany
ContactUsLinkComponent
3-Methanesulphonyloxymethylpyrrolidine-1-carboxylic acid tert-butyl ester 97%
3-Methanesulphonyloxymethylpyrrolidine-1-carboxylic acid tert-butyl ester 97%
Catalog # APOSOR912949-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methanesulphonyloxymethylpyrrolidine-1-carboxylic acid tert-butyl ester 97%
Catalog # APOSOR912949-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.36 g/mol
Storage Temperature:Ambient
CAS Number:141699-56-1
Formula:C₁₁H₂₁NO₅S
MDL Number:MFCD04038482