Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzoyl isocyanate
Benzoyl isocyanate 95%
Benzoyl isocyanate 95%
Benzoyl isocyanate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Benzoyl isocyanate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
4461-33-0
:
Formula:
C₈H₅NO₂
MDL Number:
MFCD00002032
:
147.13
Density:
1.171 g/cm³ (25 °C)
Flash Pt:
72 °C
Storage Temperature:
Freezer
UN:
2206
ADR:
6.1,III