Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methylindol-3-ylacetic acid
5-Methylindol-3-ylacetic acid 95%
5-Methylindol-3-ylacetic acid 95%
5-Methylindol-3-ylacetic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Methylindol-3-ylacetic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1912-47-6
:
Formula:
C₁₁H₁₁NO₂
MDL Number:
MFCD00022751
:
189.21
Storage Temperature:
Ambient