Order Entry
Germany
ContactUsLinkComponent
2-Isopropyl-3-methoxypyrazine 98%
2-Isopropyl-3-methoxypyrazine 98%
Catalog # APOSOR911656-25G
Supplier:  Apollo Scientific
CAS Number:  
2-Isopropyl-3-methoxypyrazine 98%
Catalog # APOSOR911656-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :152.1900 g/mol
Storage Temperature:Ambient
CAS Number:25773-40-4
Formula:C₈H₁₂N₂O
MDL Number:MFCD00006135