Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxypyrazine
2-Methoxypyrazine 98%
2-Methoxypyrazine 98%
2-Methoxypyrazine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Methoxypyrazine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
3149-28-8
:
Formula:
C₅H₆N₂O
MDL Number:
MFCD00006126
:
110.12 g/mol
Storage Temperature:
Ambient