4-Aminobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:873-74-5
:
Formula:C₇H₆N₂
MDL Number:MFCD00007821
:118.14 g/mol
Melting Pt:83…85 °C
Storage Temperature:Ambient
EINECS:212-850-1
UN:3439
ADR:6.1,III
:GHS06
:GHS07