Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methoxy-N-methylaniline hydrochloride
3-Methoxy-N-methylaniline hydrochloride 98%
3-Methoxy-N-methylaniline hydrochloride 98%
3-Methoxy-N-methylaniline hydrochloride 98%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Methoxy-N-methylaniline hydrochloride 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
26926-55-6
:
Formula:
C₈H₁₂ClNO
MDL Number:
MFCD13196469
Storage Temperature:
Ambient