4-sec-Butylaniline 98%
Catalog # APOSOR906980-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:149.24 g/mol
Storage Temperature:Ambient
CAS Number:30273-11-1
Formula:C₁₀H₁₅N
MDL Number:MFCD00010223