tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:101385-93-7
:
Formula:C₉H₁₅NO₃
MDL Number:MFCD01631194
:185.22 g/mol
Melting Pt:34…38 °C
Flash Pt:>110 °C
:GHS05
:GHS07
Storage Temperature:Refrigerator