tert-Butyl-4-methylenepiperidine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:159635-49-1
:
Formula:C₁₁H₁₉NO₂
MDL Number:MFCD01632522
Boiling Pt:75…80 °C/1mmHg
:197.2741 g/mol
:GHS07
Storage Temperature:Ambient