Methyl nicotinate 98%
Catalog # APOSOR905628-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:137.14 g/mol
Storage Temperature:Ambient
CAS Number:93-60-7
Formula:C₇H₇NO₂
MDL Number:MFCD00006388