Order Entry
Germany
Orders LinkContactUsLinkComponent
Thioisonicotinamide 98%

Thioisonicotinamide 98%

Supplier:   Apollo Scientific
Thioisonicotinamide 98%
Supplier:  Apollo Scientific
Certificates
Thioisonicotinamide 98%

6 Options Available Below

Product Details & Documents
Synonyms:  Pyridine-4-carbothioamide
  • CAS Number:2196-13-6
  • Formula:C₆H₆N₂S
  • MDL Number:MFCD00006437
  • Molecular Weight:138.19 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 31,90 €
    31,90 €
    1 * 25 g
       31,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 99,90 €
    99,90 €
    1 * 100 g
       99,90 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 329,00 €
    329,00 €
    1 * 500 g
       329,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 580,00 €
    1 580,00 €
    1 * 2,5 kg
       1 580,00 €

    Specifications:
    New Product
    New Product