tert-Butyl-4-((tosyloxy)methyl)piperidine-1-carboxylate 97%
Catalog # APOSOR901658-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:369.48 g/mol
Storage Temperature:Ambient
CAS Number:166815-96-9
Formula:C₁₈H₂₇NO₅S
MDL Number:MFCD05864740