Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Tribromoimidazole
2,4,5-Tribromoimidazole
2,4,5-Tribromoimidazole
APOSOR8896-100G
:
Apollo Scientific
:
2,4,5-Tribromoimidazole
APOSOR8896-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
304.77 g/mol
:
217…220 °C
:
Refrigerator
:
217-997-5
:
2811
:
6.1,II
:
GHS06
:
2034-22-2
:
C₃HBr₃N₂
:
MFCD00005184