Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Chlorophenol 99%

Irritant
Environment
New Product
Supplier:   Apollo Scientific
4-Chlorophenol 99%
Supplier:  Apollo Scientific
New Product
Certificates
4-Chlorophenol 99%

3 Options Available Below

Product Details & Documents
Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Boiling Pt:217…220 °C
  • Melting Pt:39…45 °C
  • Density:1.306 g/cm³ (25 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
  • EINECS:203-402-6
  • UN:2020
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 39,10 €
    39,10 €
    1 * 10 g
       39,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 85,40 €
    85,40 €
    1 * 25 g
       85,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 151,00 €
    151,00 €
    1 * 50 g
       151,00 €

    Specifications:
    New Product
    New Product