
2,4,5-Trichloropyrimidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5750-76-5
:
Formula:C₄HCl₃N₂
MDL Number:MFCD03788200
:183.42 g/mol
Boiling Pt:84 °C/1mmHg
Density:1.6 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient