Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-3,5-di-tert-butylbenzene
1-Bromo-3,5-di-tert-butylbenzene
1-Bromo-3,5-di-tert-butylbenzene
:
Apollo Scientific
1-Bromo-3,5-di-tert-butylbenzene
:
Apollo Scientific
:
22385-77-9
:
:
C₁₄H₂₁Br
:
MFCD00796945
:
269.23 g/mol
:
62…66 °C
:
GHS07
:
Ambient