

3-Chloro-2-methoxypyridine 98%
Catalog # APOSOR8560-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:143.57 g/mol
Boiling Pt:50…53 °C/3mmHg
Density:1.25 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:13472-84-9
Formula:C₆H₆ClNO
MDL Number:MFCD03095227