Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Amino-3-bromo-2-methoxypyridine
5-Amino-3-bromo-2-methoxypyridine
5-Amino-3-bromo-2-methoxypyridine
Irritant
Corrosive
5-Amino-3-bromo-2-methoxypyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Corrosive
5-Amino-3-bromo-2-methoxypyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
53242-18-5
:
Formula:
C₆H₇BrN₂O
MDL Number:
MFCD06659518
:
203.0400 g/mol
Melting Pt:
53…55 °C
:
GHS07
:
GHS05
Storage Temperature:
Ambient