Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-Thz-OH
Fmoc-Thz-OH 97%
Fmoc-Thz-OH 97%
Fmoc-Thz-OH 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Fmoc-Thz-OH 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
133054-21-4
:
Formula:
C₁₉H₁₇NO₄S
MDL Number:
MFCD00235910
:
355.408 g/mol
Melting Pt:
153…158 °C
:
GHS07
Storage Temperature:
Ambient