0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
tert-Butyl-4-carbamoylpiperidine-1-carboxylate
tert-Butyl-4-carbamoylpiperidine-1-carboxylate
tert-Butyl-4-carbamoylpiperidine-1-carboxylate
:
Apollo Scientific
tert-Butyl-4-carbamoylpiperidine-1-carboxylate
:
Apollo Scientific
:
91419-48-6
:
:
C₁₁H₂₀N₂O₃
:
MFCD02180953
:
228.2900 g/mol
:
163…164 °C
:
GHS07
:
Ambient