Order Entry
Germany
ContactUsLinkComponent
Boc-Thz-OH 98%
Boc-Thz-OH 98%
Catalog # APOSOR8031-100G
Supplier:  Apollo Scientific
CAS Number:  
Boc-Thz-OH 98%
Catalog # APOSOR8031-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :233.282 g/mol
Melting Pt:132…136 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:51077-16-8
Formula:C₉H₁₅NO₄S
MDL Number:MFCD00077002