6-Chloro-2-methylindole 98%
Catalog # APOSOR7950-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:165.62 g/mol
Melting Pt:75…77 °C
Storage Temperature:Ambient
CAS Number:6127-17-9
Formula:C₉H₈ClN
MDL Number:MFCD09027716