Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pimelic acid
Pimelic acid
Pimelic acid
Pimelic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Pimelic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
111-16-0
:
Formula:
C₇H₁₂O₄
MDL Number:
MFCD00004425
:
160.17 g/mol
Boiling Pt:
212 °C/10mmHg
Melting Pt:
103…105 °C
:
GHS07
Storage Temperature:
Ambient