Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-3-(aminomethyl)benzoate hydrochloride
Methyl-3-(aminomethyl)benzoate hydrochloride
Methyl-3-(aminomethyl)benzoate hydrochloride
:
Apollo Scientific
Methyl-3-(aminomethyl)benzoate hydrochloride
:
Apollo Scientific
:
17841-68-8
:
:
C₉H₁₂ClNO₂
:
MFCD04039173
:
201.65 g/mol
:
179…181 °C
:
GHS07
:
Ambient