Order Entry
Germany
Orders LinkContactUsLinkComponent
6-Hydroxyindole 95%

6-Hydroxyindole 95%

Corrosive
Irritant
Environment
Supplier:   Apollo Scientific
6-Hydroxyindole 95%
Supplier:  Apollo Scientific
Certificates
6-Hydroxyindole 95%

3 Options Available Below

Product Details & Documents
Synonyms:  Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol
  • CAS Number:2380-86-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152101
  • Molecular Weight:133.15 g/mol
  • Melting Pt:125…128 °C
  • Storage Temperature:Refrigerator
  • EINECS:417-020-4
  • UN:3077
  • ADR:9+EHS,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 59,40 €
    59,40 €
    1 * 1 g
       59,40 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 189,00 €
    189,00 €
    1 * 5 g
       189,00 €