Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4'-aminobiphenyl-3-carboxylate
Methyl-4'-aminobiphenyl-3-carboxylate
Methyl-4'-aminobiphenyl-3-carboxylate
Irritant
Environment
Methyl-4'-aminobiphenyl-3-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Environment
Methyl-4'-aminobiphenyl-3-carboxylate
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
400747-22-0
:
Formula:
C₁₄H₁₃NO₂
MDL Number:
MFCD04039111
Storage Temperature:
Ambient
UN:
3077
ADR:
9+EHS,III
:
GHS07
:
GHS09
Related Information
Documentation
Certificates
Search by Lot Number
Documentation