Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4'-Methoxybiphenyl-2-carboxylic acid
4'-Methoxybiphenyl-2-carboxylic acid
4'-Methoxybiphenyl-2-carboxylic acid
4'-Methoxybiphenyl-2-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
4'-Methoxybiphenyl-2-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
18110-71-9
:
Formula:
C₁₄H₁₂O₃
MDL Number:
MFCD03426469
:
228.25 g/mol
:
GHS07
Storage Temperature:
Ambient