0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1-(1-Naphthyl)-1H-pyrrole-2,5-dione
1-(1-Naphthyl)-1H-pyrrole-2,5-dione
1-(1-Naphthyl)-1H-pyrrole-2,5-dione
Irritant
1-(1-Naphthyl)-1H-pyrrole-2,5-dione
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-(1-Naphthyl)-1H-pyrrole-2,5-dione
Supplier:
Apollo Scientific
About this item
CAS Number:
3369-39-9
:
Formula:
C₁₄H₉NO₂
MDL Number:
MFCD00019715
:
223.23 g/mol
Melting Pt:
112…118 °C
:
GHS07
Storage Temperature:
Ambient