N,N,N',N'-Tetramethylethylenediamine (TEMED) 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:110-18-9
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008335
:116.21 g/mol
Boiling Pt:120…122 °C
Melting Pt:–55 °C
Density:0.775 g/cm³ (25 °C)
Flash Pt:20 °C
Storage Temperature:Ambient
EINECS:203-744-6
UN:2372
ADR:3,II
:GHS02
:GHS05
:GHS07