Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Naphtholbenzeine
1-Naphtholbenzeine
1-Naphtholbenzeine
1-Naphtholbenzeine
Supplier:
Apollo Scientific
Certificates
Expand image
1-Naphtholbenzeine
Supplier:
Apollo Scientific
About this item
CAS Number:
145-50-6
:
6948-88-5
Formula:
C₂₇H₁₈O₂
MDL Number:
MFCD00078492
:
374.44 g/mol
Melting Pt:
∼ 225 °C (dec.)
EINECS:
205-656-3
:
GHS07
Storage Temperature:
Ambient