Order Entry
Germany
ContactUsLinkComponent
1,1,3,3-Tetramethyldisilazane
1,1,3,3-Tetramethyldisilazane
Catalog # APOSOR70075-100G
Supplier:  Apollo Scientific
CAS Number:  
1,1,3,3-Tetramethyldisilazane
Catalog # APOSOR70075-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:99…100 °C
Density:0.766 g/cm³ (25 °C)
Flash Pt:–3 °C
Storage Temperature:Refrigerator
ADR:3+8,II
 :133.34 g/mol
UN:2924
 :GHS02
 :GHS05
CAS Number:15933-59-2
Formula:C₄H₁₅NSi₂
MDL Number:MFCD00025626