0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Methyl 3-bromo-1H-indazole-4-carboxylate
Methyl-3-bromo-1H-indazole-4-carboxylate
Methyl-3-bromo-1H-indazole-4-carboxylate
APOSOR70023-1G
:
Apollo Scientific
:
Methyl-3-bromo-1H-indazole-4-carboxylate
APOSOR70023-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
255.0681 g/mol
:
132…133 °C
:
GHS07
:
Ambient
:
885271-63-6
:
C₉H₇BrN₂O₂
:
MFCD03840642