Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Acetylindole-3-carboxaldehyde
1-Acetylindole-3-carboxaldehyde
1-Acetylindole-3-carboxaldehyde
:
Apollo Scientific
1-Acetylindole-3-carboxaldehyde
:
Apollo Scientific
:
22948-94-3
:
:
C₁₁H₉NO₂
:
MFCD00039691
:
187.20 g/mol
:
164…166 °C
:
GHS07
:
Ambient