Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Chlorocinnamic acid 98%

Bottle, 25 g
Catalog # APOSOR6578-25G
Irritant
Supplier:   Apollo Scientific
4-Chlorocinnamic acid 98%
Catalog # APOSOR6578-25G
Supplier:  Apollo Scientific
CAS Number:  1615-02-7
4-Chlorocinnamic acid 98%

Bottle, 25 g

CAS Number:  1615-02-7
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-(4-Chlorophenyl)acrylic acid
  • CAS Number:1615-02-7
  • Formula:C₉H₇ClO₂
  • MDL Number:MFCD00004396
  • Molecular Weight:182.61 g/mol
  • Melting Pt:248…250 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.