Order Entry
Germany
ContactUsLinkComponent
5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid
5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid
Catalog # APOSOR6268-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid
Catalog # APOSOR6268-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208.24 g/mol
Storage Temperature:Ambient
CAS Number:914637-30-2
Formula:C₉H₈N₂O₂S
MDL Number:MFCD08056306