Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl-4-formylpyridin-3-ylcarbamate
tert-Butyl-4-formylpyridin-3-ylcarbamate 97%
tert-Butyl-4-formylpyridin-3-ylcarbamate 97%
:
Apollo Scientific
tert-Butyl-4-formylpyridin-3-ylcarbamate 97%
:
Apollo Scientific
:
116026-95-0
:
:
C₁₁H₁₄N₂O₃
:
MFCD02683209
:
222.25 g/mol
:
GHS07
:
Ambient