Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl azetidin-3-ylcarbamate
tert-Butyl azetidin-3-ylcarbamate
tert-Butyl azetidin-3-ylcarbamate
tert-Butyl azetidin-3-ylcarbamate
Supplier:
Apollo Scientific
Certificates
Expand image
tert-Butyl azetidin-3-ylcarbamate
Supplier:
Apollo Scientific
About this item
CAS Number:
91188-13-5
:
Formula:
C₈H₁₆N₂O₂
MDL Number:
MFCD01861759
ADR:
9+EHS,III
:
172.23 g/mol
UN:
3077
:
GHS07
Storage Temperature:
Ambient